Ytterbium(III)trifluormethansulfonat Hydrat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Trifluoromethanesulphonic acid ytterbium salt hydrate, Ytterbium(III) trifluoromethanesulphonate hydrate, Ytterbium(III) triflate hydrate, Ytterbium triflate hydrate

M-Clarity™ Qualitätsstufe = MQ200

405329-25G
SIAL405329-25GEA 220 EUR
SIAL405329-25G
Ytterbium(III)trifluormethansulfonat Hydrat, Sigma-Aldrich®
Ytterbium(III)trifluormethansulfonat Hydrat
Formel: C₃F₉O₉S₃Yb·xH₂O
MDL: MFCD03703499
CAS-Nummer: 252976-51-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR