Lanthan(III)trifluormethansulfonat Hydrat, Sigma-Aldrich®

Lieferant: Merck

405337-5G
SIAL405337-5GEA 59.9 EUR
SIAL405337-5G
Lanthan(III)trifluormethansulfonat Hydrat, Sigma-Aldrich®
Lanthan(III)trifluormethansulfonat Hydrat
Formel: C₃F₉LaO₉S₃·H₂O
Molecular Weight: 604,13 g/mol
CAS-Nummer: 34629-21-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR