Lutetium(III)trifluormethansulfonat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Lutetium(III) trifluoromethanesulphonate

405345-1G
SIAL405345-1GEA 358 EUR
SIAL405345-1G
Lutetium(III)trifluormethansulfonat, Sigma-Aldrich®
Lutetium(III)trifluormethansulfonat
Formel: C₃F₉LuO₉S₃
MDL: MFCD00192254
CAS-Nummer: 126857-69-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR