2-Allylcyclohexanon, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

405752-5G
SIAL405752-5GEA 161 EUR
SIAL405752-5G
2-Allylcyclohexanon, Sigma-Aldrich®
2-Allylcyclohexanon
Formel: C₉H₁₄O
MDL: MFCD00043548
CAS-Nummer: 94-66-6
EINECS: 202-352-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR