2,3,4-Trimethyl-2-penten, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

406082-1G
SIAL406082-1GEA 57.5 EUR
SIAL406082-1G
2,3,4-Trimethyl-2-penten, Sigma-Aldrich®
2,3,4-Trimethyl-2-pentene
Formel: C₈H₁₆
MDL: MFCD00059214
CAS-Nummer: 565-77-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR