10-Undecinsäure, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Undec-10-ynoic acid
M-Clarity™ Qualitätsstufe = MQ200
406546-5G
SIAL406546-5GEA
95.1
EUR
SIAL406546-5G
10-Undecinsäure, Sigma-Aldrich®
10-Undecynoic acid
Formel:
C₁₁H₁₈O₂ Molecular Weight: 182,26 g/mol |
MDL:
MFCD00014389 CAS-Nummer: 2777-65-3 EINECS: 220-471-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...