Trimethylolpropane tris[poly(propylene glycol), amine terminated] ether, M.W. 440 ∼Mn, Sigma-Aldrich®

Lieferant: Merck
Gefahr


M-Clarity™ Qualitätsstufe = MQ100

406708-250ML
SIAL406708-250MLEA 115 EUR
SIAL406708-250ML
Trimethylolpropane tris[poly(propylene glycol), amine terminated] ether, M.W. 440 ∼Mn, Sigma-Aldrich®
Trimethylolpropane tris[poly(propylene glycol), amine terminated] ether
Formel: C₂H₅C[CH₂[OCH₂CH(CH₃)]nNH₂]₃
Dichte: 0,981 g/mL (25 °C) (Lit.)
Flammpunkt: 113 °C
MDL: MFCD00197951
CAS-Nummer: 39423-51-3
UN: 3082
ADR: 9,III

Bestellen

Spezifikation Testergebnisse

Appearance (Color) Colorless to Very Faint Yellow
Appearance (Form) Liquid
Infrared Spectrum Conforms to Structure
Color Test < 50 APHA
Water Content < 0.25 %
Total Amines 6.1 - 6.6 meq/g
Primary Amine 90 - 100 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR