[2-(2-Methoxyethoxy)ethoxy]essigsäure (3,6,9-Trioxadecansäure), Sigma-Aldrich®

Lieferant: Merck

Synonyme: [2-(2-Methoxyethoxy)ethoxy]acetic acid, 3,6,9-Trioxadecanoic acid, 2-(2-(2-Methoxyethoxy)ethoxy)acetic acid

M-Clarity™ Qualitätsstufe = MQ200

407003-250ML
SIAL407003-250MLEA 156 EUR
SIAL407003-250ML
[2-(2-Methoxyethoxy)ethoxy]essigsäure (3,6,9-Trioxadecansäure), Sigma-Aldrich®
[2-(2-Methoxyethoxy)ethoxy]essigsäure (3,6,9-Trioxadecansäure)
Formel: C₇H₁₄O₅
Molecular Weight: 178,1831 g/mol
MDL: MFCD00044098
CAS-Nummer: 16024-58-1
EINECS: 240-162-1
UN: 3265
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR