4-(4-Bromphenyl)-4-piperidinol, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

407445-5G
SIAL407445-5GEA 79.4 EUR
SIAL407445-5G
4-(4-Bromphenyl)-4-piperidinol, Sigma-Aldrich®
4-(4-Bromphenyl)-4-piperidinol
Formel: C₁₁H₁₄BrNO
MDL: MFCD00067034
CAS-Nummer: 57988-58-6
EINECS: 261-065-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR