2,4-Diamino-6-methyl-1,3,5-triazin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2,4-Diamino-6-methyl-s-triazin, 6-Methyl-1,3,5-triazin-2,4-diamin, 6-Methyl-1,3,5-triazin-2,4-diyldiamin

M-Clarity™ Qualitätsstufe = MQ100

407801-100G
SIAL407801-100GEA 42.3 EUR
SIAL407801-100G
2,4-Diamino-6-methyl-1,3,5-triazin, Sigma-Aldrich®
2,4-Diamino-6-methyl-1,3,5-triazine
Formel: C₄H₇N₅
Molecular Weight: 125,13 g/mol
MDL: MFCD00023192
CAS-Nummer: 542-02-9
EINECS: 208-796-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR