3-Brom-5-iodbenzoesäure, Sigma-Aldrich®

Lieferant: Merck
Achtung


M-Clarity™ Qualitätsstufe = MQ100

408085-5G
SIAL408085-5GEA 72.1 EUR
SIAL408085-5G
3-Brom-5-iodbenzoesäure, Sigma-Aldrich®
3-Brom-5-iodbenzoesäure
Formel: C₇H₄BrIO₂
Molecular Weight: 326,92 g/mol
MDL: MFCD00191851
CAS-Nummer: 188815-32-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR