N(α)-Boc-L-histidin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Boc-His-OH

M-Clarity™ Qualitätsstufe = MQ100

408433-5G
SIAL408433-5GEA 109 EUR
SIAL408433-5G
N(α)-Boc-L-histidin, Sigma-Aldrich®
N(α)-Boc-L-histidin
Formel: C₁₁H₁₇N₃O₄
Molecular Weight: 255,3 g/mol
MDL: MFCD00065576
CAS-Nummer: 17791-52-5
EINECS: 241-768-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR