1,3-Butandioldimethacrylat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1,3-Butylene glycol dimethacrylate

M-Clarity™ Qualitätsstufe = MQ100

408999-250ML
SIAL408999-250MLEA 167 EUR
SIAL408999-250ML
1,3-Butandioldimethacrylat, Sigma-Aldrich®
1,3-Butandioldimethacrylat
Formel: C₁₂H₁₈O₄
Molecular Weight: 226,27 g/mol
MDL: MFCD00014930
CAS-Nummer: 1189-08-8
EINECS: 214-711-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR