Tributylgerman, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Tributylgermanium, Tributylgermaniumhydrid, Hydridotributylgerman

409170-1G
SIAL409170-1GEA 237 EUR
SIAL409170-1G
Tributylgerman, Sigma-Aldrich®
Tributylgerman
Formel: C₁₂H₂₈Ge
Molecular Weight: 244,96 g/mol
Siedepunkt: 123 °C (20 mmHg)
Dichte: 0,949 g/cm³ (20 °C)
MDL: MFCD00077987
CAS-Nummer: 998-39-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR