1-Butyl-1-methylpyrrolidiniumbis(trifluormethylsulfonyl)imid, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

40963-5G-F
SIAL40963-5G-FEA 67 EUR
SIAL40963-5G-F
1-Butyl-1-methylpyrrolidiniumbis(trifluormethylsulfonyl)imid, Sigma-Aldrich®
1-Butyl-1-methylpyrrolidiniumbis(trifluormethylsulfonyl)imid
Formel: C₁₁H₂₀F₆N₂O₄S₂
MDL: MFCD07784447
CAS-Nummer: 223437-11-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR