5-Brom-3-methoxysalicylaldehyd, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
5-Brom-2-hydroxy-3-methoxybenzaldehyd
410470-25G
SIAL410470-25GEA
241
EUR
SIAL410470-25G
5-Brom-3-methoxysalicylaldehyd, Sigma-Aldrich®
5-Brom-3-methoxysalicylaldehyd
Formel:
C₈H₇BrO₃ Molecular Weight: 231,05 g/mol Siedepunkt: 314 °C (1013 hPa) Schmelzpunkt: 128 °C |
MDL:
MFCD00016593 CAS-Nummer: 5034-74-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...