3-Brom-3-buten-1-ol, Sigma-Aldrich®

Lieferant: Merck

410888-1G
SIAL410888-1GEA 359 EUR
SIAL410888-1G
3-Brom-3-buten-1-ol, Sigma-Aldrich®
3-Brom-3-buten-1-ol
Formel: C₄H₇BrO
Molecular Weight: 151 g/mol
MDL: MFCD00154041
CAS-Nummer: 76334-36-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR