Diethyl(bromdifluormethyl)phosphonat, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

411361-5G
SIAL411361-5GEA 96.2 EUR
SIAL411361-5G
Diethyl(bromdifluormethyl)phosphonat, Sigma-Aldrich®
Diethyl(bromdifluormethyl)phosphonat
Formel: C₅H₁₀BrF₂O₃P
Molecular Weight: 267,01 g/mol
MDL: MFCD00069118
CAS-Nummer: 65094-22-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR