3,5-Dibrombenzoesäure, Sigma-Aldrich®

Lieferant: Merck
Achtung


M-Clarity™ Qualitätsstufe = MQ100

411523-5G
SIAL411523-5GEA 134 EUR
SIAL411523-5G
3,5-Dibrombenzoesäure, Sigma-Aldrich®
3,5-Dibromobenzoic acid
Formel: C₇H₄Br₂O₂
Molecular Weight: 279,92 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00051758
CAS-Nummer: 618-58-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR