1,3-Butandioldiacrylat, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

411752-25ML
SIAL411752-25MLEA 75 EUR
SIAL411752-25ML
1,3-Butandioldiacrylat, Sigma-Aldrich®
1,3-Butandioldiacrylat
Formel: C₁₀H₁₄O₄
Molecular Weight: 198,22 g/mol
CAS-Nummer: 19485-03-1
EINECS: 243-105-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR