(Ethoxycarbonylmethyl)triphenylphosphoniumbromid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (Carbethoxymethyl)triphenylphosphonium bromide, (2-Ethoxy-2-oxoethyl)triphenylphosphonium bromide

M-Clarity™ Qualitätsstufe = MQ100

412252-100G
SIAL412252-100GEA 63.8 EUR
SIAL412252-100G
(Ethoxycarbonylmethyl)triphenylphosphoniumbromid, Sigma-Aldrich®
(Ethoxycarbonylmethyl)triphenylphosphoniumbromid
Formel: C₂₂H₂₂BrO₂P
Molecular Weight: 429.30 g/mol
MDL: MFCD00011835
CAS-Nummer: 1530-45-6
EINECS: 216-230-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR