4-(Trifluormethyl)phenylhydrazin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-Hydrazinobenzotrifluorid, 1-(4-(Trifluormethyl)phenyl)hydrazin

M-Clarity™ Qualitätsstufe = MQ200

412295-5G
SIAL412295-5GEA 78.5 EUR
SIAL412295-5G
4-(Trifluormethyl)phenylhydrazin, Sigma-Aldrich®
4-(Trifluormethyl)phenylhydrazin
Formel: C₇H₇F₃N₂
Molecular Weight: 176.14
MDL: MFCD00042508
CAS-Nummer: 368-90-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR