4-(2-Methyl-2-propanyl)-2-nitrophenol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-(tert-Butyl)-2-nitrophenol

M-Clarity™ Qualitätsstufe = MQ100

412600-25G
SIAL412600-25GEA 198 EUR
SIAL412600-25G
4-(2-Methyl-2-propanyl)-2-nitrophenol, Sigma-Aldrich®
4-tert-Butyl-2-nitrophenol
Formel: C₁₀H₁₃NO₃
Molecular Weight: 195,2151 g/mol
MDL: MFCD00191888
CAS-Nummer: 3279-07-0
EINECS: 221-914-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR