Dimethylcyclopropan-1,1-dicarboxylat, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

412783-50ML
SIAL412783-50MLEA 74.6 EUR
SIAL412783-50ML
Dimethylcyclopropan-1,1-dicarboxylat, Sigma-Aldrich®
Dimethylcyclopropan-1,1-dicarboxylat
Formel: C₇H₁₀O₄
MDL: MFCD00192079
CAS-Nummer: 6914-71-2
EINECS: 414-240-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR