2-Hydroxy-4-octyloxybenzophenon, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (2-Hydroxy-4-(octyloxy)phenyl)(phenyl)methanone

M-Clarity™ Qualitätsstufe = MQ200

413151-100G
SIAL413151-100GEA 77.8 EUR
SIAL413151-100G
2-Hydroxy-4-octyloxybenzophenon, Sigma-Aldrich®
2-Hydroxy-4-octyloxybenzophenon
Formel: C₂₁H₂₆O₃
Molecular Weight: 326,44 g/mol
MDL: MFCD00027327
CAS-Nummer: 1843-05-6
EINECS: 217-421-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR