Dioctylphenylphosphonat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Di-n-octyl phenylphosphonate

M-Clarity™ Qualitätsstufe = MQ200

413429-25ML
SIAL413429-25MLEA 187 EUR
SIAL413429-25ML
Dioctylphenylphosphonat, Sigma-Aldrich®
Dioctylphenylphosphonat
Formel: C₂₂H₃₉O₃P
Molecular Weight: 382,53 g/mol
MDL: MFCD00009561
CAS-Nummer: 1754-47-8
EINECS: 217-142-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR