1-Allyl-3-methylimidazoliumbromid, Sigma-Aldrich®

Lieferant: Merck

41382-5G
SIAL41382-5GEA 173 EUR
SIAL41382-5G
1-Allyl-3-methylimidazoliumbromid, Sigma-Aldrich®
1-Allyl-3-methylimidazoliumbromid
Formel: C₇H₁₁N₂Br
Molecular Weight: 203,08 g/mol
MDL: MFCD08277003
CAS-Nummer: 31410-07-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR