Uracil-5-carboxaldehyde 98% (durch HPLC), Sigma-Aldrich®

Lieferant: Merck
Achtung

Synonyme: 2,4-Dioxo-1,2,3,4-tetrahydropyrimidine-5-carbaldehyde, 5-Formyluracil

M-Clarity™ Qualitätsstufe = MQ100

414174-1G
SIAL414174-1GEA 125 EUR
SIAL414174-1G
Uracil-5-carboxaldehyde 98% (durch HPLC), Sigma-Aldrich®
Uracil-5-carboxaldehyde

5-Formyluracil can be used for the preparation of covalently linked base with 5-aminocytosine pair via Schiff base formation.

Formel: C₅H₄N₂O₃
Molecular Weight: 140,10 g/mol
Siedepunkt: >300 °C (Zers., Lit.)
MDL: MFCD00192185
CAS-Nummer: 1195-08-0

Bestellen

Spezifikation Testergebnisse

Appearance (Color) White to Off-White
Appearance (Form) Powder or Crystalline Powder
Infrared Spectrum Conforms to Structure
Carbon 41.8 - 43.9%
Nitrogen 19.5 - 20.5%
Purity (HPLC) ≥97.5%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR