1-Iod-2-(trimethylsilyl)acetylen, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (Iodoethynyl)trimethylsilane

M-Clarity™ Qualitätsstufe = MQ100

414670-1G
SIAL414670-1GEA 46 EUR
SIAL414670-1G
1-Iod-2-(trimethylsilyl)acetylen, Sigma-Aldrich®
1-Iod-2-(trimethylsilyl)acetylen
Formel: C₅H₉ISi
Molecular Weight: 224,12 g/mol
MDL: MFCD00274201
CAS-Nummer: 18163-47-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR