5-Iod-1,3-dimethyluracil, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

415294-1G
SIAL415294-1GEA 0 EUR
SIAL415294-1G
5-Iod-1,3-dimethyluracil, Sigma-Aldrich®
5-Iod-1,3-dimethyluracil
Formel: C₆H₇IN₂O₂
Molecular Weight: 266,04 g/mol
MDL: MFCD00192228
CAS-Nummer: 40738-83-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR