Bis(pentamethylcyclopentadienyl)mangan(II), Sigma-Aldrich®

Lieferant: Merck

Synonyme: Decamethylmanganocene, Bis(pentamethylcyclopentadienyl)manganese

M-Clarity™ Qualitätsstufe = MQ100

415405-1G
SIAL415405-1GEA 141 EUR
SIAL415405-1G
Bis(pentamethylcyclopentadienyl)mangan(II), Sigma-Aldrich®
Bis(pentamethylcyclopentadienyl)mangan(II)
Formel: C₂₀H₃₀Mn
Molecular Weight: 325,39 g/mol
MDL: MFCD00058844
CAS-Nummer: 67506-86-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR