1,2-Dichlor-1,1,2,2-tetramethyldisilan, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1,2-Dichlorotetramethyldisilane

M-Clarity™ Qualitätsstufe = MQ100

415456-5ML
SIAL415456-5MLEA 98.6 EUR
SIAL415456-5ML
1,2-Dichlor-1,1,2,2-tetramethyldisilan, Sigma-Aldrich®
1,2-Dichlor-1,1,2,2-tetramethyldisilan
Formel: C₄H₁₂Cl₂Si₂
MDL: MFCD00078528
CAS-Nummer: 4342-61-4
EINECS: 224-400-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR