(R)-(+)-3-Aminochinuclidin Dihydrochlorid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (R)-3-Aminoquinuclidine dihydrochloride

M-Clarity™ Qualitätsstufe = MQ200

415715-1G
SIAL415715-1GEA 46.3 EUR
SIAL415715-1G
(R)-(+)-3-Aminochinuclidin Dihydrochlorid, Sigma-Aldrich®
(R)-(+)-3-Aminochinuclidin Dihydrochlorid
Formel: C₇H₁₆Cl₂N₂
Molecular Weight: 199,1213 g/mol
MDL: MFCD00191752
CAS-Nummer: 123536-14-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR