(S)-(-)-3-Aminochinuclidin Dihydrochlorid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (S)-Quinuclidin-3-amine dihydrochloride, (S)-Quinuclidin-3-amine 2HCl, (S)-(-)-3-Amino-1-azabicyclo[2.2.2]octane dihydrochloride

M-Clarity™ Qualitätsstufe = MQ100

415723-1G
SIAL415723-1GEA 65.8 EUR
SIAL415723-1G
(S)-(-)-3-Aminochinuclidin Dihydrochlorid, Sigma-Aldrich®
(S)-(-)-3-Aminochinuclidin Dihydrochlorid
Formel: C₇H₁₆Cl₂N₂
Molecular Weight: 199,1213 g/mol
MDL: MFCD00191753
CAS-Nummer: 119904-90-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR