2'-Hydroxy-5'-methyl-3'-nitroacetophenon, Sigma-Aldrich®

Lieferant: Merck
Achtung


M-Clarity™ Qualitätsstufe = MQ100

415766-5G
SIAL415766-5GEA 40 EUR
SIAL415766-5G
2'-Hydroxy-5'-methyl-3'-nitroacetophenon, Sigma-Aldrich®
2'-Hydroxy-5'-methyl-3'-nitroacetophenon
Formel: C₉H₉NO₄
Molecular Weight: 195,17 g/mol
MDL: MFCD00192216
CAS-Nummer: 66108-30-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR