5'-Chlor-2'-hydroxy-3'-nitroacetophenon, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1-Chlor-3-ethanoyl-4-hydroxy-5-nitrobenzene, 1-(5-Chloro-2-hydroxy-3-nitrophenyl)ethanone

M-Clarity™ Qualitätsstufe = MQ100

415774-25G
SIAL415774-25GEA 38.9 EUR
SIAL415774-25G
5'-Chlor-2'-hydroxy-3'-nitroacetophenon, Sigma-Aldrich®
5'-Chlor-2'-hydroxy-3'-nitroacetophenon
Formel: C₈H₆ClNO₄
Molecular Weight: 215,59 g/mol
MDL: MFCD00191921
CAS-Nummer: 84942-40-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR