2-(3-Brompropoxy)tetrahydropyran, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

415979-5G
SIAL415979-5GEA 179 EUR
SIAL415979-5G
2-(3-Brompropoxy)tetrahydropyran, Sigma-Aldrich®
2-(3-Brompropoxy)tetrahydropyran
Formel: C₈H₁₅BrO₂
MDL: MFCD00058593
CAS-Nummer: 33821-94-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR