S-tert-Butylthioacetat, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ200

416738-1G
SIAL416738-1GEA 90.3 EUR
SIAL416738-1G
S-tert-Butylthioacetat, Sigma-Aldrich®
S-tert-Butylthioacetat
Formel: C₆H₁₂OS
Molecular Weight: 132,23 g/mol
CAS-Nummer: 999-90-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR