1,1,1-Trifluor-3-phenylaceton, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3-Phenyl-1,1,1-trifluoropropan-2-one, 1,1,1-Trifluoro-3-phenyl-2-propanone, (3,3,3-Trifluor-2-propanoyl)benzene, 1,1,1-Trifluoro-3-phenylpropan-2-one

M-Clarity™ Qualitätsstufe = MQ100

416789-5G
SIAL416789-5GEA 126 EUR
SIAL416789-5G
1,1,1-Trifluor-3-phenylaceton, Sigma-Aldrich®
1,1,1-Trifluor-3-phenylaceton
Formel: C₉H₇F₃O
Molecular Weight: 188,15 g/mol
MDL: MFCD00068171
CAS-Nummer: 350-92-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR