Triethyl-2-phosphonobutyrat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Ethyl 2-(diethoxyphosphoryl)butanoate

417467-5ML
SIAL417467-5MLEA 60.6 EUR
SIAL417467-5ML
Triethyl-2-phosphonobutyrat, Sigma-Aldrich®
Triethyl-2-phosphonobutyrat
Formel: C₁₀H₂₁O₅P
Molecular Weight: 252,25 g/mol
MDL: MFCD00041347
CAS-Nummer: 17145-91-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR