6-Methylchromon Hydrat ≥98%, Sigma-Aldrich®

Lieferant: Merck
Achtung


M-Clarity™ Qualitätsstufe = MQ100

417475-1G
SIAL417475-1GEA 52.7 EUR
SIAL417475-1G
6-Methylchromon Hydrat ≥98%, Sigma-Aldrich®
6-Methylchromon Hydrat

6-Methylchromone hydrate is a chromone derivative. Chromones are cyclic compounds containing nine carbon atoms with sp2 hybridization. They are commonly referred as 1,4-benzapyranes or γ-benzopyrones.


Application
6-Methylchromone may be employed as activated alkene in the methanolic trimethylamine or sodium methoxide catalyzed Baylis-Hillman coupling reaction with aromatic and aliphatic aldehydes.

Formel: C₁₀H₈O₂·xH₂O
Molecular Weight: 160,17 g/mol (anhy)
MDL: MFCD00209598
CAS-Nummer: 207511-19-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR