Bis(2,2,6,6-tetramethyl-3,5-heptandionato-O,O')zink(II), Sigma-Aldrich®

Lieferant: Merck

Synonyme: Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)zinc(II), Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)zinc, Zn(TMHD)2

417734-1G
SIAL417734-1GEA 87 EUR
SIAL417734-1G
Bis(2,2,6,6-tetramethyl-3,5-heptandionato-O,O')zink(II), Sigma-Aldrich®
Bis(2,2,6,6-tetramethyl-3,5-heptandionato-O,O')zink(II)
Formel: C₂₂H₃₈O₄Zn
Molecular Weight: 431,92 g/mol
MDL: MFCD00192578
CAS-Nummer: 14363-14-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR