tert-Butylhydroperoxid 5-6 mol/l in n-Nonan, Sigma-Aldrich®
Lieferant: Merck
|
Gefahr
|
Synonyme:
TBHP
M-Clarity™ Qualitätsstufe = MQ200
418064-50ML
SIAL418064-50MLEA
171
EUR
SIAL418064-50ML
tert-Butylhydroperoxid 5-6 mol/l in n-Nonan, Sigma-Aldrich®
tert-Butylhydroperoxid
Formel:
C₄H₁₀O₂ Molecular Weight: 90.12 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD00002130 CAS-Nummer: 75-91-2 EINECS: 200-915-7 UN: 3109 ADR: 5.2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...