Bariumtrifluormethansulfonat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Barium Bis(trifluoromethanesulphonate), Barium triflate, Barium(II) trifluoromethanesulphonate, Trifluoromethanesulphonic acid Barium(II) salt, Ba(OTf)2, Barium(II) triflate

419087-5G
SIAL419087-5GEA 76.6 EUR
SIAL419087-5G
Bariumtrifluormethansulfonat, Sigma-Aldrich®
Bariumtrifluormethansulfonat
Formel: C₂BaF₆O₆S₂
Molecular Weight: 435.45 g/mol
MDL: MFCD00143644
CAS-Nummer: 2794-60-7
EINECS: 220-526-6
UN: 1564
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR