2,5-Di-tert-butyl-p-benzochinon, Sigma-Aldrich®

Lieferant: Merck

419648-5G
SIAL419648-5GEA 58 EUR
SIAL419648-5G
2,5-Di-tert-butyl-p-benzochinon, Sigma-Aldrich®
2,5-Di-tert-butyl-p-benzochinon
Formel: C₁₄H₂₀O₂
MDL: MFCD00019442
CAS-Nummer: 2460-77-7
EINECS: 219-552-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR