6-Methoxyflavon, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 6-Methoxy-2-phenyl-4H-chromen-4-on, 6-Methoxy-2-phenyl-4H-1-benzopyran-4-on, 6-Methoxy-2-phenyl-4-benzopyron, 6-Methoxy-2-phenylchromon, 6-Methoxy-2-phenylchromen-4-on, 6-Methoxy-2-phenyl-4H-benzo[b]pyran-4-on

M-Clarity™ Qualitätsstufe = MQ100

419737-1G
SIAL419737-1GEA 104 EUR
SIAL419737-1G
6-Methoxyflavon, Sigma-Aldrich®
6-Methoxyflavon
Formel: C₁₆H₁₂O₃
Molecular Weight: 252,27 g/mol
MDL: MFCD00017322
CAS-Nummer: 26964-24-9
EINECS: 248-144-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR