4'-Formylbenzo-15-krone-5 (4'-Formylbenzo-1,4,7,10,13-pentaoxacyclopentadecan), Sigma-Aldrich®
Lieferant: Merck
Synonyme:
4'-Formylbenzo-15-crown 5-Ether, 2,3-(4-Formylbenzo)-1,4,7,10,13-pentaoxacyclopentadec-2-ene, 2,5,8,11,14-Pentaoxabicyclo[13.4.0]nonadeca-1(15),16,18-triene-17-carbaldehyde
M-Clarity™ Qualitätsstufe = MQ200
419974-250MG
SIAL419974-250MGEA
417
EUR
SIAL419974-250MG
4'-Formylbenzo-15-krone-5 (4'-Formylbenzo-1,4,7,10,13-pentaoxacyclopentadecan), Sigma-Aldrich®
4'-Formylbenzo-15-krone-5 (4'-Formylbenzo-1,4,7,10,13-pentaoxacyclopentadecan)
Formel:
C₁₅H₂₀O₆ Molecular Weight: 296.32 g/mol |
MDL:
MFCD00192180 CAS-Nummer: 60835-73-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...