Siliciumdioxid 30% in Wasser, kolloidale Dispersion, LUDOX® SM, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Silicium(IV)oxid

M-Clarity™ Qualitätsstufe = MQ100

420794-1L 420794-4L
SIAL420794-1LEA 54.7 EUR
SIAL420794-1L SIAL420794-4L
Siliciumdioxid 30% in Wasser, kolloidale Dispersion, LUDOX® SM, Sigma-Aldrich®
Siliciumdioxid
Formel: SiO₂
Molecular Weight: 60.08 g/mol
Siedepunkt: 2230
Schmelzpunkt: 1719
Dichte: 1.22 g/ml (25 °C)
MDL: MFCD00011232
CAS-Nummer: 7631-86-9
Merck Index: 14,08493

Bestellen

Spezifikation Testergebnisse

Colour Colourless or White
Form Liquid
ICP Major Analysis (Silica component) Confirmed
Specific gravity at 60 °F 1,209 - 1,227
pH at 25 °C 9,3 - 10,3
Viscosity at 25 °C 4,8 - 6,8 cps
Silica 29,0 - 31,0%
Ratio of SiO₂/Na₂O 45 - 56
Size (Surface area) 320 - 400 (m²/g)
Sulphate (as Na₂SO₄) ≤0,06%
Transmission ≥74,0%
Concentration 30 wt% suspension in H₂O


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR