(S)-(-)-1,2,3,4-Tetrahydroisochinolin-3-carbonsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (3S)-1,2,3,4-Tetrahydro-3-isoquinolinecarbocylic acid, (S)-1,2,3,4-Tetrahydroisoquinoline-3-carboxylic acid

M-Clarity™ Qualitätsstufe = MQ100

421626-5G
SIAL421626-5GEA 23.6 EUR
SIAL421626-5G
(S)-(-)-1,2,3,4-Tetrahydroisochinolin-3-carbonsäure, Sigma-Aldrich®
(S)-(-)-1,2,3,4-Tetrahydroisochinolin-3-carbonsäure
Formel: C₁₀H₁₁NO₂
Molecular Weight: 177,20 g/mol
MDL: MFCD00144533
CAS-Nummer: 74163-81-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR