N-Cbz-L-phenylalaninol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Z-Phenylalaninol, (S)-2-(Cbz-amino)-3-phenyl-1-propanol, N-Benzyloxycarbonyl-L-phenylalaninol

M-Clarity™ Qualitätsstufe = MQ100

421723-5G
SIAL421723-5GEA 32.3 EUR
SIAL421723-5G
N-Cbz-L-phenylalaninol, Sigma-Aldrich®
N-Cbz-L-phenylalaninol
Formel: C₁₇H₁₉NO₃
Molecular Weight: 285,3 g/mol
MDL: MFCD00191138
CAS-Nummer: 6372-14-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR